missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-(3,4-dihydroxyphenyl)ethanone, TRC

Product Code. 17943155
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943155 100 g 100g
17981145 1 g 1g
17983055 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943155 Supplier Toronto Research Chemicals Supplier No. TRCC365325100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-40-1
Molecular Formula C8 H7 Cl O3
Molecular Weight (g/mol) 186.59
Synonym α-Chloro-3,4-dihydroxyacetophenone, 1-Chloroacetyl-3,4-dihydroxybenzene, 2-Chloro-1-(3,4-dihydroxyphenyl)-1-ethanone, 2-Chloro-1-(3,4-dihydroxyphenyl)ethanone, 2-Chloro-3',4'-dihydroxyacetophenone, 3,4-Dihydroxy-α-chloroacetophenone, 3,4-Dihydroxyphenacyl chloride, 4-(Chloroacetyl)catechol, NSC 13905, NSC 17869, α-Chloro-3',4'-dihydroxyacetophenone, Noradrenaline Imp. E (EP)
IUPAC Name 2-chloro-1-(3,4-dihydroxyphenyl)ethanone
SMILES Oc1ccc(cc1O)C(=O)CCl

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H7ClO3/c9-4-8(12)5-1-2-6(10)7(11)3-5/h1-3,10-11H,4H2
Formula Weight 186.0084
Chemical Name or Material 2-Chloro-1-(3,4-dihydroxyphenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.