missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3,2-dioxaphospholane-2-oxide, TRC

Product Code. 30361849
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361849 Supplier Toronto Research Chemicals Supplier No. TRCC36575525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-1,3,2-dioxaphospholane-2-oxide
CAS 6609-64-9
Molecular Formula C2H4ClO3P
Quantity 25 g
Synonym Ethylene Phosphorochloridate, Phosphorochloridic Acid, Cyclic Ethylene Ester, 2-Chloro-2-oxido-1,3,2-dioxaphospholane, 2-Chloro-2-oxo-1,3,2-dioxaphospholane, Cyclic Ethylene Phosphorochloridate, Ethylene Chlorophosphate
InChI Formula InChI=1S/C2H4ClO3P/c3-7(4)5-1-2-6-7/h1-2H2
SMILES ClP1(=O)OCCO1
IUPAC Name 2-chloro-1,3,2λ^{5}-dioxaphospholane 2-oxide
Molecular Weight (g/mol) 142.48
Formula Weight 141.9587
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.