missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3,2-dioxaphospholane-2-oxide (Technical Grade), TRC

Product Code. 30367571
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367571 1 g -
30357576 10 g -
30362298 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367571 Supplier Toronto Research Chemicals Supplier No. TRCC3657581G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6609-64-9
Molecular Formula C2H4ClO3P
Molecular Weight (g/mol) 142.48
Synonym Ethylene Phosphorochloridate, Phosphorochloridic Acid, Cyclic Ethylene Ester, 2-Chloro-2-oxido-1,3,2-dioxaphospholane, 2-Chloro-2-oxo-1,3,2-dioxaphospholane, Cyclic Ethylene Phosphorochloridate, Ethylene Chlorophosphate
IUPAC Name 2-chloro-1,3,2λ^{5}-dioxaphospholane 2-oxide
SMILES ClP1(=O)OCCO1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H4ClO3P/c3-7(4)5-1-2-6-7/h1-2H2
Formula Weight 141.9587
Chemical Name or Material 2-Chloro-1,3,2-dioxaphospholane-2-oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.