missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%

Product Code. 10217353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217353 100 g Glass bottle 100g
10286011 5 g Glass bottle 5g
10245531 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10217353 Supplier Thermo Scientific Acros Supplier No. 332061000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6609-64-9 | C2H4ClO3P | 142.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6609-64-9
Molecular Formula C2H4ClO3P
Molecular Weight (g/mol) 142.48
MDL Number MFCD00043138
InChI Key SBMUNILHNJLMBF-UHFFFAOYSA-N
Synonym 2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate
PubChem CID 81087
IUPAC Name 2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
SMILES C1COP(=O)(O1)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 11.0°C to 14.0°C
Color White
Density 1.5500g/mL
Boiling Point 98.0°C to 99.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4540
Quantity 100 g
Specific Gravity 1.55
Formula Weight 142.48
Percent Purity 95%
Physical Form Solid Film
Chemical Name or Material 2-Chloro-1, 3, 2-dioxaphospholane-2-oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.