missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-{1H,2H,3H,4H,5H-pyrido[4,3-b]indol-2-yl}ethan-1-one, TRC

Product Code. 30345768
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345768 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345768 Supplier Toronto Research Chemicals Supplier No. TRCB418838500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-1-(1H,2H,3H,4H,5H-pyrido[4,3-b]indol-2-yl)ethan-1-one
CAS 1087784-16-4
Molecular Formula C13H13N2OCl
Quantity 500 mg
InChI Formula InChI=1S/C13H13ClN2O/c14-7-13(17)16-6-5-12-10(8-16)9-3-1-2-4-11(9)15-12/h1-4,15H,5-8H2
SMILES ClCC(=O)N1CCc2[nH]c3ccccc3c2C1
IUPAC Name 2-chloro-1-(1,3,4,5-tetrahydropyrido[4,3-b]indol-2-yl)ethanone
Molecular Weight (g/mol) 248.71
Formula Weight 248.0716
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.