missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-1,1-dimethoxy-Ethane, TRC

Product Code. 30337472
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30337472 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337472 Supplier Toronto Research Chemicals Supplier No. TRCC0680105G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-chloro-1,1-dimethoxy-Ethane
CAS 97-97-2
Molecular Formula C4H9ClO2
Quantity 5 g
Synonym chloro-Acetaldehyde dimethyl acetal, 2-Chloro-1,1-dimethoxyethane, 1,1-Dimethoxy-2-chloroethane, 1-Chloro-2,2-dimethoxyethane, 2,2-Dimethoxyethyl chloride, 2-Chloroacetaldehyde dimethyl acetal, Chloroacetaldehyde dimethyl acetal, NSC 60388
InChI Formula InChI=1S/C4H9ClO2/c1-6-4(3-5)7-2/h4H,3H2,1-2H3
SMILES COC(CCl)OC
IUPAC Name 2-chloro-1,1-dimethoxyethane
Molecular Weight (g/mol) 124.57
Formula Weight 124.0291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.