missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether, 97%

Product Code. p-7046899
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339365 5 g 5g
11349365 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 11339365 Supplier Thermo Scientific Alfa Aesar Supplier No. L16725.06
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 13838-16-9 | C3H2ClF5O | 184.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13838-16-9
Molecular Formula C3H2ClF5O
Molecular Weight (g/mol) 184.49
MDL Number MFCD00069095
InChI Key JPGQOUSTVILISH-UHFFFAOYNA-N
Synonym enflurane, methylflurether, efrane, ethrane, 2-chloro-1-difluoromethoxy-1,1,2-trifluoroethane, alyrane, anesthetic 347, 2-chloro-1,1,2-trifluoroethyl difluoromethyl ether, compound 347, anesthetic compound no. 347
PubChem CID 3226
ChEBI CHEBI:4792
IUPAC Name 2-chloro-1-(difluoromethoxy)-1,1,2-trifluoroethane
SMILES FC(F)OC(F)(F)C(F)Cl

Specifications

Density 1.517
Boiling Point 56°C to 57°C
Flash Point None
Linear Formula CHF2OCF2CHClF
Refractive Index 1.303
Quantity 5 g
Beilstein 1737129
Merck Index 14,3581
Formula Weight 184.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.