missing translation for 'onlineSavingsMsg'
Learn More

(2-Carboxyethyl)triphenylphosphonium chloride, 98%

Product Code. 11451637
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451637 25 g 25g
11441637 5 g 5g
11461637 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451637 Supplier Thermo Scientific Alfa Aesar Supplier No. B25609.14
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 36626-29-6 | C21H20ClO2P | 370.81 g/mol

2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36626-29-6
Molecular Formula C21H20ClO2P
Molecular Weight (g/mol) 370.81
MDL Number MFCD00185663
InChI Key GALLWJZTZYJVSL-UHFFFAOYSA-N
Synonym 2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride
PubChem CID 10270534
SMILES [Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 197°C to 201°C
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in dimethyl sulfoxide and methanol.
IUPAC Name (2-carboxyethyl)triphenylphosphanium chloride
Formula Weight 370.81
Percent Purity 98%
Chemical Name or Material (2-Carboxyethyl)triphenylphosphonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.