Learn More
2-Carboxybenzaldehyde, 98+%
Description
2-Carboxybenzaldehyde is a carboxylated derivative of benzaldehyde which is readily converted to 2-hydroxymethyl benzoic acid by CBA dehydrogenase. 2-Carboxybenzaldehyde is a metabolite of ampicillin phthalidyl ester. It has been used to synthesize a series of N-substituted isoindolinones by reductive C-N coupling and intramolecular amidation with amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Carboxybenzaldehyde is a carboxylated derivative of benzaldehyde which is readily converted to 2-hydroxymethyl benzoic acid by CBA dehydrogenase. 2-Carboxybenzaldehyde is a metabolite of ampicillin phthalidyl ester. It has been used to synthesize a series of N-substituted isoindolinones by reductive C-N coupling and intramolecular amidation with amines.
Solubility
Soluble in water.
Notes
Air Sensitive. Store away from air and oxidizing agents. Incompatible with strong bases.
Specifications
Specifications
| Melting Point | 97°C to 101°C |
| Density | 1.404 |
| Flash Point | 198°C (388°F) |
| Quantity | 1000 g |
| Beilstein | 742381 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 150.13 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2-Carboxybenzaldehyde |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.