missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxy-4-chlorobenzeneboronic acid, 96%, Thermo Scientific™

Catalog Number 15439817
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
850568-07-9
C7H6BClO4
200.381
MFCD06659869
RENDEFHQQIYNJL-UHFFFAOYSA-N
2-carboxy-4-chlorophenylboronic acid, 5-chloro-2-dihydroxyboranyl benzoic acid, 2-carboxy-4-chlorobenzeneboronic acid, 2-borono-5-chlorobenzoicacid, 2-carboxy-4-chloro benzeneboronic acid, benzoic acid,2-borono-5-chloro-9ci, 5-chloranyl-2-dihydroxyboranyl benzoic acid, 5-chloro-2-dihydroxyboryl benzoic acid
23005367
2-borono-5-chlorobenzoic acid
B(C1=C(C=C(C=C1)Cl)C(=O)O)(O)O
This item is not returnable. View return policy
850568-07-9
C7H6BClO4
200.381
MFCD06659869
RENDEFHQQIYNJL-UHFFFAOYSA-N
2-carboxy-4-chlorophenylboronic acid, 5-chloro-2-dihydroxyboranyl benzoic acid, 2-carboxy-4-chlorobenzeneboronic acid, 2-borono-5-chlorobenzoicacid, 2-carboxy-4-chloro benzeneboronic acid, benzoic acid,2-borono-5-chloro-9ci, 5-chloranyl-2-dihydroxyboranyl benzoic acid, 5-chloro-2-dihydroxyboryl benzoic acid
23005367
2-borono-5-chlorobenzoic acid
B(C1=C(C=C(C=C1)Cl)C(=O)O)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 250 mg
Formula Weight 200.39
Percent Purity 96%
Chemical Name or Material 2-Carboxy-4-chlorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'