missing translation for 'onlineSavingsMsg'
Learn More

2-Carbamoylbenzeneboronic acid, 96%

Product Code. 15439397
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439397 250 mg 250mg
15429397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439397 Supplier Thermo Scientific Alfa Aesar Supplier No. H52642.MD
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 380430-54-6 | C7H8BNO3 | 164.955 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-54-6
Molecular Formula C7H8BNO3
Molecular Weight (g/mol) 164.955
MDL Number MFCD02179454
InChI Key LBWJTKOVBMVJJX-UHFFFAOYSA-N
Synonym 2-aminocarbonylphenylboronic acid, 2-carbamoylphenyl boronic acid, 2-carbamoylbenzeneboronic acid, 2-aminocarbonyl benzeneboronic acid, 2-boronobenzamide, boronic acid, 2-aminocarbonyl phenyl, 2-aminocarbonylphenyl boronic acid, 2-aminocarbonyl phenylboronic acid, boronic acid,b-2-aminocarbonyl phenyl, pubchem1766
PubChem CID 2737809
IUPAC Name (2-carbamoylphenyl)boronic acid
SMILES B(C1=CC=CC=C1C(=O)N)(O)O

Specifications

Melting Point 226°C to 230°C
Quantity 250 mg
Formula Weight 164.96
Percent Purity 96%
Chemical Name or Material 2-Carbamoylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.