missing translation for 'onlineSavingsMsg'
Learn More

2-Butyn-1-ol, 98%

Product Code. 11457677
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457677 5 g 5g
11467677 25 g 25g
11477677 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457677 Supplier Thermo Scientific Alfa Aesar Supplier No. A15709.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 764-01-2 | C4H6O | 70.091 g/mol

2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

Solubility
Miscible with chloroform and methanol.

Notes
Incompatible with oxidizing agents, acids, acid anhydrides and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 764-01-2
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00002914
InChI Key NEEDEQSZOUAJMU-UHFFFAOYSA-N
Synonym 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
PubChem CID 12991
IUPAC Name but-2-yn-1-ol
SMILES CC#CCO

Specifications

Melting Point -2°C
Density 0.938
Boiling Point 142°C to 143°C
Flash Point 51°C (123°F)
Odor Characteristic
Refractive Index 1.453
Quantity 5 g
UN Number UN1987
Beilstein 1733676
Solubility Information Miscible with chloroform and methanol.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material 2-Butyn-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Butyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: But-2-yn-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.