missing translation for 'onlineSavingsMsg'
Learn More

2-Butyn-1-ol, 98%

Product Code. 10308650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308650 25 mL Glass bottle 25mL
10348600 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10308650 Supplier Thermo Scientific Acros Supplier No. 223400250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 764-01-2 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 764-01-2
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00002914
InChI Key NEEDEQSZOUAJMU-UHFFFAOYSA-N
Synonym 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
PubChem CID 12991
IUPAC Name but-2-yn-1-ol
SMILES CC#CCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2.2°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 142.0°C to 143.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C≡CCH2OH
Refractive Index 1.4520 to 1.4540
Quantity 25 mL
Beilstein 01, II, 505
Specific Gravity 0.93
Formula Weight 70.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butyn-1-ol, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Butyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.