missing translation for 'onlineSavingsMsg'
Learn More

2-Butyn-1-al diethyl acetal, 98%, Thermo Scientific™

Product Code. 10328650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10328650 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10328650 Supplier Thermo Scientific Acros Supplier No. 223410010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2806-97-5
CAS Min % 97.5
CAS Max % 100.0
Density 0.9000g/mL
Boiling Point 65.0°C to 67.0°C (12.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H14O2
Linear Formula CH3C≡CCH(OC2H5)2
Refractive Index 1.4250 to 1.4270
MDL Number MFCD00009274
Quantity 1g
Beilstein 01, 750
Fieser 15,66
Specific Gravity 0.9
Synonym 2-butynal diethyl acetal, 2-butyn-1-al diethyl acetal, 2-butynyl aldehyde diethyl acetal, 2-butyne, 1,1-diethoxy, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, acmc-20ao12, tetrolaldehyde diethyl acetal, 1,1-diethoxy-2-butyne #
InChI Key FKKVKKSEVMQYER-UHFFFAOYSA-N
SMILES CCOC(OCC)C#CC
IUPAC Name 1,1-diethoxybut-2-yne
Molecular Weight (g/mol) 142.20
PubChem CID 137721
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material 2-Butyn-1-al diethyl acetal, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Butyn-1-al diethyl acetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.