missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butylfuran 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2-Butylfuran |
| CAS | 4466-24-4 |
| Analyte or Component Names | 2-Butylfuran |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H12 O |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-n-Butylfuran, Furan, 2-butyl- |
| InChI Formula | InChI=1S/C8H12O/c1-2-3-5-8-6-4-7-9-8/h4,6-7H,2-3,5H2,1H3 |
| SMILES | CCCCc1occc1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction