missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid (Losartan Carboxylic Acid), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid (Losartan Carboxylic Acid)
Specifications
Specifications
| Chemical Name or Material | Losartan Carboxylic Acid |
| CAS | 124750-92-1 |
| Analyte or Component Names | Losartan Carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H21 Cl N6 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid, E 3174, EXP 3174, Losartan Acid, 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid |
| InChI Formula | InChI=1S/C22H21ClN6O2/c1-2-3-8-18-24-20(23)19(22(30)31)29(18)13-14-9-11-15(12-10-14)16-6-4-5-7-17(16)21-25-27-28-26-21/h4-7,9-12H,2-3,8,13H2,1H3,(H,30,31)(H,25,26,27,28) |
| SMILES | CCCCc1nc(Cl)c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4 |
| IUPAC Name | 2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction