missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-1H-imidazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10223232
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10223232 10 g 10g
10079451 1 g 1g
10314062 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10223232 Supplier Thermo Scientific Maybridge Supplier No. SB02028EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 68282-49-5
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD01075030
InChI Key PTHGVOCFAZSNNA-UHFFFAOYSA-N
Synonym 2-butyl-1h-imidazole-4-carbaldehyde, 2-butyl-5-formylimidazole, 2-butyl-4-formylimidazole, 2-butyl-4-imidazolyl formaldehyde, unii-u12g73sqsx, 2-butyl-1h-imidazole-5-carboxaldehyde, u12g73sqsx, 1h-imidazole-5-carboxaldehyde, 2-butyl, 2-butylimidazole-4-carbaldehyde, 2-butyl-3h-imidazole-4-carbaldehyde
PubChem CID 2735671
IUPAC Name 2-butyl-1H-imidazole-5-carbaldehyde
SMILES CCCCC1=NC=C(N1)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 152.2
Percent Purity 97%
Chemical Name or Material 2-Butyl-1H-imidazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.