missing translation for 'onlineSavingsMsg'
Learn More

(2-Butyl-1H-imidazol-4-yl)methanol, 97%, Thermo Scientific™

Product Code. 10039491
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10039491 10 g 10g
10029481 1 g 1g
10082404 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10039491 Supplier Thermo Scientific Maybridge Supplier No. SB02033EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 68283-19-2
Molecular Formula C8H14N2O
Molecular Weight (g/mol) 154.21
MDL Number MFCD00239516
InChI Key UZKBZGAMRJRWLR-UHFFFAOYSA-N
Synonym 2-butyl-1h-imidazol-4-yl methanol, 2-butyl-5-hydroxymethylimidazole, 2-butyl-1h-imidazol-5-yl methanol, unii-k14mm5ob9a, 2-butyl-4-hydroxymethyl imidazole, k14mm5ob9a, 2-butylimidazol-5-yl methan-1-ol, 2-butyl-3h-imidazol-4-yl methanol, 2-n-butyl-4-imidazolemethanol, 2-n-butyl-4-hydroxymethylimidazole
PubChem CID 2735673
IUPAC Name (2-butyl-1H-imidazol-5-yl)methanol
SMILES CCCCC1=NC=C(CO)N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material (2-Butyl-1H-imidazol-4-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.