missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butoxyethyl Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Butoxyethylacetate |
| CAS | 112-07-2 |
| Molecular Formula | C8 H16 O3 |
| Quantity | 1 mL |
| Synonym | Ethanol, 2-butoxy-, acetate (6CI,7CI,8CI,9CI), 2-Butoxyethyl acetate, B 0700, BCA, BMGAC, Butoxyethyl acetate, Butyl cellosolve acetate, Butyl glycol acetate, Dowanol EB acetate, EB Acetate, Ethylene glycol acetate monobutyl ether, Ethylene glycol butyl ether acetate, Ethylene glycol monobutyl ether acetate, Glycol monobutyl ether acetate |
| InChI Formula | InChI=1S/C8H16O3/c1-3-4-5-10-6-7-11-8(2)9/h3-7H2,1-2H3 |
| SMILES | CCCCOCCOC(=O)C |
| IUPAC Name | 2-butoxyethyl acetate |
| Molecular Weight (g/mol) | 160.21 |
| Formula Weight | 160.1099 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?