missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethanol, TRC

Product Code. 30003850
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003850 10 mL 10mL
30004427 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003850 Supplier Toronto Research Chemicals Supplier No. TRCB69289510ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6 H14 O2
Molecular Weight (g/mol) 118.17
Synonym Ethanol, 2-butoxy- (8CI, 9CI, ACI), 2-Butoxyethanol (ACI), 2-Butoxy-1-ethanol, 2-n-Butoxyethanol, 3-Oxa-1-heptanol, A 13919, BCS, Bikanol B 1, Buchiseru, Butyl Cellosolve, Butyl Cellu-Sol, Butyl glycol, Butyl Glysolv, Butyl icinol, Butyl monoether glycol, Butyl Oxitol, C4E1, Chimec NR, Dabco PM 300, DB solvent, Dowanol EB, Eastman EB, EGBE, Ektasolve EB, Ethylene glycol butyl ether, Ethylene glycol mono-n-butyl ether, Ethylene glycol monobutyl ether, Ethylene glycol n-butyl ether, Gafcol EB, Glycol butyl ether, Glycol EB, Glycol monobutyl ether, K Foam Lo, Mearcell 3532, Minex BDH, Monobutyl glycol ether, Monoethyleneglycol butyl ether, n-Butyl cellosolve, NSC 60759, O-Butyl ethylene glycol, Poly-Solv EB, R 1171, SG, Simple Green, β-Butoxyethanol
IUPAC Name 2-butoxyethanol
SMILES CCCCOCCO

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3
Formula Weight 118.0994
Chemical Name or Material Ethyleneglycol Monobutyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.