missing translation for 'onlineSavingsMsg'
Learn More

2-Butenylmagnesium chloride, 0.5M in MeTHF, Thermo Scientific Chemicals

Product Code. 15498057
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498057 100 mL 100mL
15408067 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498057 Supplier Thermo Scientific Alfa Aesar Supplier No. H51171.AE
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6088-88-6 | Molecular Formula: C4H7ClMg | Molecular Weight: 114.855 g/mol

2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6088-88-6
Molecular Formula C4H7ClMg
MDL Number MFCD01321226
Synonym 2-butenylmagnesium chloride, 2-butenylmagnesiumchloride,0.5minmethf, 2-butenylmagnesium chloride, 0.5m in methf, 2e-but-2-en-1-yl chloro magnesium, 2-butenylmagnesium chloride solution, 0.5 m in thf
PubChem CID 6104506

Specifications

Linear Formula CH3CH=CHCH2MgCl
Quantity 100 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information It reacts violently with water.
InChI Key XQAMFOKVRXJOGD-CZEFNJPISA-M
SMILES CC=C[CH2-].[Mg+2].[Cl-]
IUPAC Name magnesium;(E)-but-2-ene;chloride
Molecular Weight (g/mol) 114.855
Formula Weight 114.86
Concentration or Composition (by Analyte or Components) 0.5M in MeTHF
Chemical Name or Material 2-Butenylmagnesium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.