missing translation for 'onlineSavingsMsg'
Learn More

2-Butanoylsulfanylethyl(trimethyl)azanium Chloride, TRC

Product Code. 30240572
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240572 500 mg 500mg
30240674 100 mg 100mg
30240990 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240572 Supplier Toronto Research Chemicals Supplier No. TRCB872370500MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22026-63-7
Molecular Formula C9H20ClNOS
Molecular Weight (g/mol) 225.78
Synonym S-Butyrylthiocholine Chloride
IUPAC Name 2-butanoylsulfanylethyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCC(=O)SCC[N+](C)(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H20NOS.ClH/c1-5-6-9(11)12-8-7-10(2,3)4;/h5-8H2,1-4H3;1H/q+1;/p-1
Formula Weight 225.0954
Chemical Name or Material 2-Butanoylsulfanylethyl(trimethyl)azanium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.