missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotoluene, 99%

Product Code. 11405073
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405073 25 g 25g
11415073 100 g 100g
11425073 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405073 Supplier Thermo Scientific Alfa Aesar Supplier No. A12315.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-46-5 | C7H7Br | 171.04 g/mol

2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluene is used in manufacturing pharmaceuticals (especially for nonsteroidal antiinflammatory drugs; Meclofenamic acid, Mefenamic acid) and other organic compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluen
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-46-5
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000068
InChI Key QSSXJPIWXQTSIX-UHFFFAOYSA-N
Synonym 2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
PubChem CID 7236
IUPAC Name 1-bromo-2-methylbenzene
SMILES CC1=CC=CC=C1Br

Specifications

Melting Point -27°C
Density 1.42
Boiling Point 179°C to 181°C
Flash Point 78°C (172°F)
Refractive Index 1.556
Quantity 25 g
Beilstein 1904176
Merck Index 14,1439
Solubility Information Soluble in alcohol,ether,acetone,and ethanol. Insoluble in water.
Formula Weight 171.04
Percent Purity 99%
Chemical Name or Material 2-Bromotoluene
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: o-Bromotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.