missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophenol, 97%

Product Code. 10350644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10350644 50 g Glass bottle 50g
10358690 1 g Glass bottle 1g
10541354 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10350644 Supplier Thermo Scientific Acros Supplier No. 221980500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6320-02-1 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6320-02-1
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004827
InChI Key YUQUNWNSQDULTI-UHFFFAOYSA-N
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
PubChem CID 80599
SMILES SC1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6040g/mL
Boiling Point 114°C to 116°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4SH
Refractive Index 1.6345 to 1.6365
Quantity 50 g
Beilstein 06, II, 300
Specific Gravity 1.604
Solubility Information Solubility in water: immiscible
IUPAC Name 2-chlorobenzenethiol
Formula Weight 189.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromothiophenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.