missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene, TRC

Product Code. 30338547
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338547 Supplier Toronto Research Chemicals Supplier No. TRCB6881855G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 2-bromothiophene
CAS 1003-09-4
Molecular Formula C4 H3 Br S
Quantity 5 g
Synonym 2-Thienyl Bromide, NSC 4456
InChI Formula InChI=1S/C4H3BrS/c5-4-2-1-3-6-4/h1-3H
SMILES Brc1cccs1
IUPAC Name 2-bromothiophene
Molecular Weight (g/mol) 163.036
Formula Weight 161.914
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.