missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene, TRC

Product Code. 30340882
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30340882 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30340882

Brand: Toronto Research Chemicals TRCB6881851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-bromothiophene
CAS 1003-09-4
Molecular Formula C4 H3 Br S
Quantity 1 g
Synonym 2-Thienyl Bromide, NSC 4456
InChI Formula InChI=1S/C4H3BrS/c5-4-2-1-3-6-4/h1-3H
SMILES Brc1cccs1
IUPAC Name 2-bromothiophene
Molecular Weight (g/mol) 163.036
Formula Weight 161.914
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.