missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene, 98%

Product Code. 10071830 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 10071830

Brand: Thermo Scientific Acros 154292500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1003-09-4 | C4H3BrS | 163.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-09-4
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005417
InChI Key TUCRZHGAIRVWTI-UHFFFAOYSA-N
Synonym 2-thienyl bromide, thiophene, 2-bromo, thiophene, bromo, bromothiophene, 2-bromo-thiophene, 2-bromo thiophene, thienyl bromide, 5-bromothiophene, 5-bromo-thiophene, 2-bromanylthiophene
PubChem CID 13851
IUPAC Name 2-bromothiophene
SMILES BrC1=CC=CS1

Specifications

Melting Point -10.0°C
Color Brown to Colorless
Density 1.6800g/mL
Boiling Point 149.0°C to 151.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5850 to 1.5870
Quantity 250 mL
Beilstein 17,33
Specific Gravity 1.68
Solubility Information Solubility in water: immiscible. Other solubilities: very soluble in ether, acetone, soluble in carbontetrachloride
Formula Weight 163.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromothiophene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromothiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.