missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene, 98+%

Product Code. 11433974
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433974 250 g 250g
11423974 50 g 50g
11413974 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433974 Supplier Thermo Scientific Alfa Aesar Supplier No. A11959.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1003-09-4 | C4H3BrS | 163.03 g/mol

2-Bromothiophene has been used in electrochemical reduction of a number of mono- and dihalothiophenes at carbon cathodes in dimethylformamide containing tetramethylammonium per chlorate by cyclic voltammetry and controlled-potential electrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromothiophene has been used in electrochemical reduction of a number of mono- and dihalothiophenes at carbon cathodes in dimethylformamide containing tetramethylammonium per chlorate by cyclic voltammetry and controlled-potential electrolysis.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1003-09-4
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005417
InChI Key TUCRZHGAIRVWTI-UHFFFAOYSA-N
Synonym 2-thienyl bromide, thiophene, 2-bromo, thiophene, bromo, bromothiophene, 2-bromo-thiophene, 2-bromo thiophene, thienyl bromide, 5-bromothiophene, 5-bromo-thiophene, 2-bromanylthiophene
PubChem CID 13851
IUPAC Name 2-bromothiophene
SMILES BrC1=CC=CS1

Tekniske data

Melting Point <-10°C
Density 1.701
Boiling Point 149°C to 152°C
Flash Point 58°C (136°F)
Refractive Index 1.586
Quantity 250 g
UN Number UN2929
Beilstein 104663
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 163.04
Percent Purity ≥98%
Chemical Name or Material 2-Bromothiophene
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Faresætning
  • H226-Flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H318-Causes serious eye damage.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.