missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene-3-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10387634
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1860-99-7
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.042
MDL Number MFCD09025880
InChI Key MMEGVQIGIBCTHI-UHFFFAOYSA-N
PubChem CID 12245921
IUPAC Name 2-bromothiophene-3-carbaldehyde
SMILES C1=CSC(=C1C=O)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-bromo-3-thiophenecarboxaldehyde, 3-thiophenecarboxaldehyde, 2-bromo, 2-bromo-3-formylthiophene, 2-bromo-3-thiophenecarbaldehyde, 2-bromo-thiophene-3-carbaldehyde, 2-bromothiophene-3-carboxaldehyde, 3-thiophenecarboxaldehyde,2-bromo, 2-bromo-3-thiophenaldehyde, 2-bromanylthiophene-3-carbaldehyde
Formula Weight 191.05
Percent Purity 97%
Chemical Name or Material 2-Bromothiophene-3-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'