Learn More
2-Bromothioanisole, 98%, ACROS Organics™
£15.70 - £41.80
Chemical Identifiers
CAS | 19614-16-5 |
---|---|
Molecular Formula | C7H7BrS |
Molecular Weight (g/mol) | 203.097 |
MDL Number | MFCD00000066 |
InChI Key | ALAQDUSTXPEHMH-UHFFFAOYSA-N |
Synonym | 2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene |
PubChem CID | 88166 |
IUPAC Name | 1-bromo-2-methylsulfanylbenzene |
SMILES | CSC1=CC=CC=C1Br |
Description
Chemical Identifiers
19614-16-5 | |
203.097 | |
ALAQDUSTXPEHMH-UHFFFAOYSA-N | |
88166 | |
CSC1=CC=CC=C1Br |
C7H7BrS | |
MFCD00000066 | |
2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene | |
1-bromo-2-methylsulfanylbenzene |
Specifications
19614-16-5 | |
100.0 | |
Authentic | |
C7H7BrS | |
1.6320 to 1.6340 (20°C, 589nm) | |
2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene | |
CSC1=CC=CC=C1Br | |
203.097 | |
203.11 | |
Liquid |
97.5 | |
Colorless to Yellow | |
98% | |
BrC6H4SCH3 | |
MFCD00000066 | |
ALAQDUSTXPEHMH-UHFFFAOYSA-N | |
1-bromo-2-methylsulfanylbenzene | |
88166 | |
≥97.5% | |
2-Bromothioanisole |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning