missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole, 99%

Product Code. 11432607
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432607 10 g 10g
11319659 25 g 25g
11442607 50 g 50g
11452607 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11432607 Supplier Thermo Scientific Alfa Aesar Supplier No. A14838.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3034-53-5 | C3H2BrNS | 164.02 g/mol

2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-53-5
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD00005316
InChI Key RXNZFHIEDZEUQM-UHFFFAOYSA-N
Synonym 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
PubChem CID 76430
IUPAC Name 2-bromo-1,3-thiazole
SMILES C1=CSC(=N1)Br

Specifications

Density 1.836
Boiling Point 171°C
Flash Point 63°C (145°F)
Refractive Index 1.593
Quantity 10 g
Beilstein 105724
Solubility Information It is insoluble in water.
Formula Weight 164.02
Percent Purity 99%
Chemical Name or Material 2-Bromothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.