missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoterephthalic acid, 97%

Product Code. 11476997
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11476997

Brand: Thermo Scientific Alfa Aesar A14393.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-35-6 | C8H5BrO4 | 245.028 g/mol

2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-35-6
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002403
InChI Key QPBGNSFASPVGTP-UHFFFAOYSA-N
Synonym bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9
PubChem CID 68513
IUPAC Name 2-bromoterephthalic acid
SMILES C1=CC(=C(C=C1C(=O)O)Br)C(=O)O

Specifications

Melting Point 297°C to 300°C
Quantity 25 g
UN Number UN2811
Beilstein 2616163
Solubility Information Soluble in water.
Formula Weight 245.03
Percent Purity 97%
Chemical Name or Material 2-Bromoterephthalic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.