missing translation for 'onlineSavingsMsg'
Learn More

2-Bromostyrene, 96%, stab. with ca 0.05% 4-tert-butylcatechol

Product Code. 11441217
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441217 25 g 25g
11431217 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441217 Supplier Thermo Scientific Alfa Aesar Supplier No. B24181.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2039-88-5 | C8H7Br | 183.048 g/mol

2-Bromostyrene is used as a cross linking agent in fire-resistant bromostyrene-crosslinked polyesters. Further, it is used to prepare trans-1-(2-bromophenyl)-2-(3-bromophenyl)ethene by reacting with 1-bromo-3-iodo-benzene, involving palladium(II) acetate as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromostyrene is used as a cross linking agent in fire-resistant bromostyrene-crosslinked polyesters. Further, it is used to prepare trans-1-(2-bromophenyl)-2-(3-bromophenyl)ethene by reacting with 1-bromo-3-iodo-benzene, involving palladium(II) acetate as catalyst.

Solubility
Miscible with methanol.

Notes
Incompatible with acids, bases, oxidizing agents and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-88-5
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
MDL Number MFCD00000076
InChI Key SSZOCHFYWWVSAI-UHFFFAOYSA-N
Synonym 2-bromostyrene, 1-bromo-2-vinylbenzene, o-bromostyrene, benzene, 1-bromo-2-ethenyl, unii-9a96gp6z5d, styrene, o-bromo, 2-bromo-styrene, o-bromovinylbenzene, pubchem23975, 2-bromo-1-vinylbenzene
PubChem CID 16264
IUPAC Name 1-bromo-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Br

Specifications

Melting Point -53°C
Density 1.416
Boiling Point 102°C to 104°C (22 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.592
Quantity 25 g
Beilstein 2323537
Solubility Information Miscible with methanol.
Formula Weight 183.05
Percent Purity 96%
Chemical Name or Material 2-Bromostyrene, stabilized with ≈0.05% 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.