missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyrimidine, 98+%

Product Code. 11329659
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329659 1 g 1g
11454864 5 g 5g
11464864 25 g 25g
11474864 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11329659 Supplier Thermo Scientific Alfa Aesar Supplier No. A13791.03
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 4595-60-2 | C4H3BrN2 | 158.99 g/mol

2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4595-60-2
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.99
MDL Number MFCD00014601
InChI Key PGFIHORVILKHIA-UHFFFAOYSA-N
Synonym pyrimidine, 2-bromo, 2-brompyrimidin, bromopyrimidine, 2-bromo pyrimidine, 2-bromo-pyrimidine, 2-bromanylpyrimidine, pubchem6901, acmc-1aqvo, 2-bromo-1,3-diazine, 2-bromopyrimidine
PubChem CID 78345
IUPAC Name 2-bromopyrimidine
SMILES BrC1=NC=CC=N1

Specifications

Melting Point 53°C to 57°C
Boiling Point 62°C to 64°C (1.5 mmHg)
Quantity 1 g
Beilstein 605945
Solubility Information Miscible with water.
Formula Weight 158.99
Percent Purity ≥98%
Chemical Name or Material 2-Bromopyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.