missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-5-carboxaldehyde, 95%

Product Code. 15430809
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430809 25 g 25g
15410809 1 g 1g
15420809 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430809 Supplier Thermo Scientific Alfa Aesar Supplier No. H63867.14
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149806-06-4 | C6H4BrNO | 186.01 g/mol

A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. 2-7°C Refrigerate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149806-06-4
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.01
MDL Number MFCD04115419
InChI Key PVUKGNBRJFTFNJ-UHFFFAOYSA-N
Synonym 6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
PubChem CID 11769234
IUPAC Name 6-bromopyridine-3-carbaldehyde
SMILES BrC1=CC=C(C=O)C=N1

Specifications

Melting Point 104°C to 110°C (decomposition)
Color Yellow
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol.
Formula Weight 186.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromopyridine-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.