missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-4-carboxamide, 97%

Product Code. 11399845
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399845 5 g 5g
11389845 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399845 Supplier Thermo Scientific Alfa Aesar Supplier No. L19854.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29840-73-1 | C6H5BrN2O | 201.023 g/mol

2-Bromopyridine-4-carboxamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyridine-4-carboxamide is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29840-73-1
Molecular Formula C6H5BrN2O
Molecular Weight (g/mol) 201.023
MDL Number MFCD00234156
InChI Key KCELTDZFDHPTBI-UHFFFAOYSA-N
PubChem CID 4625407
IUPAC Name 2-bromopyridine-4-carboxamide
SMILES C1=CN=C(C=C1C(=O)N)Br

Specifications

Melting Point 192°C
Quantity 5 g
Beilstein 471892
Synonym 2-bromoisonicotinamide, 2-bromo-isonicotinamide, 2-bromo-4-pyridinecarboxamide, 2-bromopyridin-4-ylcarboxamide, 4-pyridinecarboxamide, 2-bromo, 2-bromo-isonicotimide, 4-pyridinecarboxamide,2-bromo, pubchem16641, acmc-1cr70, 2-bromanylpyridine-4-carboxamide
Formula Weight 201.02
Percent Purity 97%
Chemical Name or Material 2-Bromopyridine-4-carboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.