missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropiophenone, 96%

Product Code. 11433414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433414 100 g 100g
11423414 25 g 25g
11443414 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433414 Supplier Thermo Scientific Alfa Aesar Supplier No. A10661.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2114-00-3 | C9H9BrO | 213.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2114-00-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00000145
InChI Key WPDWOCRJBPXJFM-UHFFFAOYSA-N
Synonym 2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
PubChem CID 16452
IUPAC Name 2-bromo-1-phenylpropan-1-one
SMILES CC(C(=O)C1=CC=CC=C1)Br

Specifications

Density 1.417
Boiling Point 245°C to 250°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.572
Quantity 100 g
UN Number UN2927
Beilstein 508550
Formula Weight 213.08
Percent Purity 96%
Chemical Name or Material 2-Bromopropiophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.