missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropene, 99%, stab.

Product Code. 11319065
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319065 5 g 5g
11329065 25 g 25g
11339065 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11319065 Supplier Thermo Scientific Alfa Aesar Supplier No. L14229.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 557-93-7 | C3H5Br | 120.977 g/mol

2-Bromopropene is used as a solvent and as an intermediate for organic synthesis. It is also used as a fumigant. Further, it reacts with nonanal to prepare 2-methyl-undec-1-en-3-ol using chromium(II) chloride as a reagent. It is utilized as a refrigerant used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropene is used as a solvent and as an intermediate for organic synthesis. It is also used as a fumigant. Further, it reacts with nonanal to prepare 2-methyl-undec-1-en-3-ol using chromium(II) chloride as a reagent. It is utilized as a refrigerant used in organic synthesis.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-93-7
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00000140
InChI Key PHMRPWPDDRGGGF-UHFFFAOYSA-N
Synonym 2-bromopropene, isopropenyl bromide, 1-propene, 2-bromo, 2-bromo-1-propene, 2-bromopropylene, propene, 2-bromo, isopropylene bromide, alpha-methylvinyl bromide, .alpha.-methylvinyl bromide, 2-bromo-propene
PubChem CID 11202
IUPAC Name 2-bromoprop-1-ene
SMILES CC(=C)Br

Specifications

Melting Point -87°C
Density 1.362
Boiling Point 47°C to 49°C
Flash Point 4°C (39°F)
Odor Pungent
Refractive Index 1.443
Quantity 5 g
UN Number UN1993
Beilstein 1731926
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 120.98
Percent Purity 99%
Chemical Name or Material 2-Bromopropene, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.