missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropane, 99%

Product Code. 11439783
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439783 1000 mL 1000mL
11449783 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439783 Supplier Thermo Scientific Alfa Aesar Supplier No. A12944.AU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-26-3 | C3H7Br | 122.993 g/mol

2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water and acetone.

Notes
Avoid all possible sources of ignition.Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-26-3
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.993
MDL Number MFCD00000147
InChI Key NAMYKGVDVNBCFQ-UHFFFAOYSA-N
PubChem CID 6358
IUPAC Name 2-bromopropane
SMILES CC(C)Br

Specifications

Melting Point -89°C
Density 1.31
Boiling Point 59°C to 60°C
Flash Point 19°C (66°F)
Assay Percent Range 99%
Linear Formula CH3CHBrCH3
Refractive Index 1.4255
Quantity 1000 mL
UN Number UN2344
Beilstein 741852
Merck Index 14,5210
Synonym isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan
Solubility Information Miscible with alcohol,benzene,chloroform and ether. Slightly miscible with water and acetone.
Formula Weight 123
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromopropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.