missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromophenylethynyl)trimethylsilane, 98%

Product Code. 15430178
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity Förpackningsstorlek
15430178 25 g 25g
15410178 1 g 1g
15420178 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 15430178 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H53402.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38274-16-7 | C11H13BrSi | 253.21 g/mol

(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38274-16-7
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.21
MDL Number MFCD01321391
InChI Key FABNGXSCLHXUOH-UHFFFAOYSA-N
Synonym 2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl
PubChem CID 4547398
SMILES C[Si](C)(C)C#CC1=CC=CC=C1Br

Specifications

Boiling Point 60°C to 80°C (2mmHg)
Quantity 25 g
Flash Point >113°C (235°F)
Refractive Index 1.554
Density 1.19
Solubility Information Slightly soluble in water.
IUPAC Name 2-(2-bromophenyl)ethynyl-trimethylsilane
Formula Weight 253.21
Percent Purity 98%
Chemical Name or Material (2-Bromophenylethynyl)trimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.