missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetone, 99%

Product Code. 10110132 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10110132 1 g Glass bottle 1g
10068761 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10110132 Supplier Thermo Scientific Acros Supplier No. 350590010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21906-31-0 | C9H9BrO | 213.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21906-31-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.08
MDL Number MFCD03410431
InChI Key TZIAZLUAMDLDJF-UHFFFAOYSA-N
Synonym 2-bromophenylacetone, 1-2-bromophenyl propan-2-one, 2-bromophenyl acetone, 1-2-bromophenyl-2-propanone, 1-acetonyl-2-bromobenzene, 2-propanone, 1-2-bromophenyl, o-bromo benyl methyl ketone, 1-2-bromophenyl acetone, o-bromo benzyl methyl ketone, pubchem23388
PubChem CID 2734092
IUPAC Name 1-(2-bromophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5575 to 1.5595
Quantity 1 g
Formula Weight 213.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromophenylacetone, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.