missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetic acid, 98+%

Product Code. 11498716
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498716 25 g 25g
11488716 5 g 5g
11408726 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498716 Supplier Thermo Scientific Alfa Aesar Supplier No. A16900.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18698-97-0 | C8H7BrO2 | 215.046 g/mol

Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

Solubility
Very faint turbidity in methanol.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18698-97-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00004314
InChI Key DWXSYDKEWORWBT-UHFFFAOYSA-N
Synonym 2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
PubChem CID 87754
IUPAC Name 2-(2-bromophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Br

Specifications

Melting Point 105°C to 106°C
Quantity 25 g
Beilstein 2250655
Solubility Information Very faint turbidity in methanol.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 2-Bromophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.