missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenyl acetate, 98%, Thermo Scientific™

Product Code. 15266404 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15266404 1 g 1g
15229816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15266404 Supplier Thermo Scientific Acros Supplier No. 452750010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1829-37-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
InChI Key BEHBHYYPTOHUHX-UHFFFAOYSA-N
Synonym o-bromophenyl acetate, 2-bromophenol acetate, 1-acetoxy-2-bromobenzene, 2-bromophenyl acetate, o-acetoxybromobenzene, phenol, 2-bromo-, acetate, phenol, o-bromo-, acetate, pubchem3055, 2-bromo-phenol acetate
PubChem CID 74586
IUPAC Name (2-bromophenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1Br

Specifications

Assay Percent Range 97.5 % min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 2-Bromophenyl acetate
compliance-icons
Product Identifier
  • 2-Bromophenyl acetate, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.