missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenol, TRC

Product Code. 30341914
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341914 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341914 Supplier Toronto Research Chemicals Supplier No. TRCB68616550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromophenol
CAS 95-56-7
Molecular Formula C6 H5 Br O
Quantity 50 g
Synonym Phenol, o-bromo- (8CI), 2-Bromophenol, 2-Hydroxy-1-bromobenzene, NSC 6970, o-Bromophenol
InChI Formula InChI=1S/C6H5BrO/c7-5-3-1-2-4-6(5)8/h1-4,8H
SMILES Oc1ccccc1Br
IUPAC Name 2-bromophenol
Molecular Weight (g/mol) 173.01
Formula Weight 171.9524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.