missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenol, 98%

Product Code. p-76467 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10357520

Brand: Thermo Scientific Acros 157360100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-56-7 | C6H5BrO | 173.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-56-7
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002146
InChI Key VADKRMSMGWJZCF-UHFFFAOYSA-N
Synonym o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
PubChem CID 7244
IUPAC Name 2-bromophenol
SMILES C1=CC=C(C(=C1)O)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5°C
Color Colorless to Yellow
Density 1.4900g/mL
Boiling Point 195°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OH
Refractive Index 1.5880 to 1.59
Quantity 10 mL
Beilstein 06, 197
Merck Index 15, 1439
Specific Gravity 1.49
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with chloroform and ether
Formula Weight 173.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromophenol, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.