missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopentane, tech. 90%

Product Code. 11423427
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423427 500 g 500g
11403427 25 g 25g
11413427 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423427 Supplier Thermo Scientific Alfa Aesar Supplier No. A17205.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-81-3 | C5H11Br | 151.047 g/mol

Used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chemical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-81-3
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.047
MDL Number MFCD00000160
InChI Key LGAJYTCRJPCZRJ-UHFFFAOYSA-N
Synonym pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
PubChem CID 7890
IUPAC Name 2-bromopentane
SMILES CCCC(C)Br

Specifications

Melting Point -96°C
Density 1.22
Boiling Point 117°C to 118°C
Flash Point 20°C (68°F)
Odor Characteristic
Refractive Index 1.4403
Quantity 500 g
UN Number UN2343
Beilstein 1718829
Solubility Information Not miscible or difficult to mix in water. Soluble in Dichloromethane,Ether,Ethyl Acetate,Hexane and Methanol
Formula Weight 151.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Bromopentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.