missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopentane, 90%, tech.

Product Code. 10070570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10070570 500 mL Glass bottle 500mL
10766321 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10070570 Supplier Thermo Scientific Acros Supplier No. 107235000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 107-81-3 | C5H11Br | 151.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-81-3
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000160
InChI Key LGAJYTCRJPCZRJ-UHFFFAOYSA-N
Synonym pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
PubChem CID 7890
IUPAC Name 2-bromopentane
SMILES CCCC(C)Br

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point -95.5°C
Density 1.2200g/mL
Boiling Point 116.0°C to 117.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 12% max. (3-Bromopentane) Typical (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH(Br)CH3
Refractive Index 1.4393 to 1.4413
Quantity 500 mL
Beilstein 01, 131
Specific Gravity 1.22
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with organic solvents
Formula Weight 151.05
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2-Bromopentane, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromopentane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.