missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)naphthalene, 96%

Product Code. 10343912 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10343912 25 g Glass bottle 25g
10722011 1 g Glass bottle 1g
10521092 5 g Glass bottle 5g
10222752 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10343912 Supplier Thermo Scientific Acros Supplier No. 162560250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 939-26-4 | C11H9Br | 221.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-26-4
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00004123
InChI Key RUHJZSZTSCSTCC-UHFFFAOYSA-N
Synonym 2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide
PubChem CID 70320
IUPAC Name 2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 52°C to 57°C
Color Yellow to Beige
Boiling Point 213°C (100.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH2Br
Quantity 25 g
Beilstein 05,568
Formula Weight 221.1
Percent Purity 96%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-(Bromomethyl)naphthalene
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Bromomethyl)naphthalene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.