missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)benzeneboronic acid, 97%

Product Code. 15489207
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489207 1 g 1g
15409217 250 mg 250mg
15499207 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15489207 Supplier Thermo Scientific Alfa Aesar Supplier No. H52467.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91983-14-1 | C7H8BBrO2 | 214.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91983-14-1
Molecular Formula C7H8BBrO2
Molecular Weight (g/mol) 214.85
MDL Number MFCD01318941
InChI Key MYVJCOQGXCONPE-UHFFFAOYSA-N
Synonym 2-bromomethylphenylboronic acid, 2-bromomethyl benzeneboronic acid, 2-bromomethyl phenylboronic acid, 2-bromomethyl phenyl boronic acid, 2-bromomethylphenylboronicacid, 2-boronobenzyl bromide, o-boronobenzyl bromide, 2-bromomethylphenyl boronic acid, pubchem7781
PubChem CID 2773278
IUPAC Name [2-(bromomethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1CBr

Specifications

Melting Point 145°C to 148°C
Quantity 1 g
Formula Weight 214.85
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.