missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-6-methylpyridine, 97%

Product Code. 15421678
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421678 5 g 5g
15431678 250 mg 250mg
15411678 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421678 Supplier Thermo Scientific Alfa Aesar Supplier No. H55438.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68470-59-7 | C7H8BrN | 186.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68470-59-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD04967229
InChI Key WJFDCFHWFHCLIW-UHFFFAOYSA-N
Synonym 2-bromomethyl-6-methylpyridine, pyridine, 2-bromomethyl-6-methyl, 6-methyl-2-bromomethyl pyridine, 2-bromomethyl-6-methyl-pyridine, 6-bromomethyl-2-methylpyridine, pubchem22577, acmc-209ur7, 2-methyl-6-bromomethyl-pyridine, 2-bromomethyl-6-methyl pyridine
PubChem CID 12543733
IUPAC Name 2-(bromomethyl)-6-methylpyridine
SMILES CC1=NC(CBr)=CC=C1

Specifications

Melting Point 38°C to 42°C
Quantity 5 g
Sensitivity Air sensitive
Formula Weight 186.05
Percent Purity 97%
Chemical Name or Material 2-Bromomethyl-6-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.